N-(4-Aminobenzyl)-1-methylpiperidin-4-amine dihydrochloride
Lieferant: Apollo Scientific
OR307772-1G
APOSOR307772-1GEA
433
EUR
APOSOR307772-1G
N-(4-Aminobenzyl)-1-methylpiperidin-4-amine dihydrochloride
N-(4-Aminobenzyl)-1-methylpiperidin-4-amine dihydrochloride
Formel:
C₁₃H₂₁N₃·2ClH Molecular Weight: 292,248 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12674811 CAS-Nummer: 1204810-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...