3-(4-aminobenzylamino)pyrrolidine-1-carboxylic acid tert-butyl ester hydrochloride

Lieferant: Apollo Scientific

OR307773-1G
APOSOR307773-1GEA 484 EUR
APOSOR307773-1G
3-(4-aminobenzylamino)pyrrolidine-1-carboxylic acid tert-butyl ester hydrochloride
3-(4-aminobenzylamino)pyrrolidine-1-carboxylic acid tert-butyl ester hydrochloride
Formel: C₁₆H₂₅N₃O₂·ClH
Molecular Weight: 327,85 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375307
CAS-Nummer: 1266226-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR