4-(4-amino-benzylamino)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

OR307774-1G
APOSOR307774-1GEA 467 EUR
APOSOR307774-1G
4-(4-amino-benzylamino)-piperidine-1-carboxylic acid tert-butyl ester
4-(4-amino-benzylamino)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₇N₃O₂
Molecular Weight: 305,415 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674812
CAS-Nummer: 1189106-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR