4-[(4-Amino-benzylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
OR307775-1G
APOSOR307775-1GEA
451
EUR
APOSOR307775-1G
4-[(4-Amino-benzylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
4-[(4-Amino-benzylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₈H₂₉N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12674813 CAS-Nummer: 1189105-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...