3-[(4-Amino-benzylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
OR307777-1G
APOSOR307777-1GEA
499
EUR
APOSOR307777-1G
3-[(4-Amino-benzylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
3-[(4-Amino-benzylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₈H₂₉N₃O₂ Molecular Weight: 319,442 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12674815 CAS-Nummer: 1189107-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...