4-[2-(4-Methyl-piperazin-1-yl)-ethyl]-phenylamine
Lieferant: Apollo Scientific
OR307779-1G
APOSOR307779-1GEA
0
EUR
APOSOR307779-1G
4-[2-(4-Methyl-piperazin-1-yl)-ethyl]-phenylamine
4-[2-(4-Methyl-piperazin-1-yl)-ethyl]-phenylamine
Formel:
C₁₃H₂₁N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09884759 CAS-Nummer: 262368-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...