4-(4-Aminophenethyl)piperazine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
OR307780-1G
APOSOR307780-1GEA
393
EUR
APOSOR307780-1G
4-(4-Aminophenethyl)piperazine-1-carboxylic acid tert-butyl ester
4-(4-Aminophenethyl)piperazine-1-carboxylic acid tert-butyl ester
Formel:
C₁₇H₂₇N₃O₂ Molecular Weight: 305,415 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11590790 CAS-Nummer: 329004-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...