3-[(4-Amino-phenylamino)-pyrrolidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

OR307782-1G
APOSOR307782-1GEA 459 EUR
APOSOR307782-1G
3-[(4-Amino-phenylamino)-pyrrolidine-1-carboxylic acid tert-butyl ester
3-[(4-Amino-phenylamino)-pyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₅H₂₃N₃O₂
Molecular Weight: 277,362 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026436
CAS-Nummer: 1159976-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR