4-(4-Amino-phenylamino)- piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
OR307783-1G
APOSOR307783-1GEA
467
EUR
APOSOR307783-1G
4-(4-Amino-phenylamino)- piperidine-1-carboxylic acid tert-butyl ester
4-(4-Amino-phenylamino)- piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₆H₂₅N₃O₂ Molecular Weight: 291,389 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026437 CAS-Nummer: 1071932-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...