4-[(4-Amino-phenylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
OR307784-1G
APOSOR307784-1GEA
451
EUR
APOSOR307784-1G
4-[(4-Amino-phenylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
4-[(4-Amino-phenylamino)-methyl]-piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₇H₂₇N₃O₂ Molecular Weight: 305,415 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026438 CAS-Nummer: 1159976-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...