3-(4-Amino-phenylamino)- piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

OR307785-1G
APOSOR307785-1GEA 491 EUR
APOSOR307785-1G
3-(4-Amino-phenylamino)- piperidine-1-carboxylic acid tert-butyl ester
3-(4-Amino-phenylamino)- piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₆H₂₅N₃O₂
Molecular Weight: 291.389 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026439
CAS-Nummer: 1159976-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR