3-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

OR307786-1G
APOSOR307786-1GEA 491 EUR
APOSOR307786-1G
3-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester
3-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₇N₃O₂
Molecular Weight: 305,415 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026440
CAS-Nummer: 1159976-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR