2-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
OR307787-1G
APOSOR307787-1GEA
516
EUR
APOSOR307787-1G
2-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester
2-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₁₇H₂₇N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026441 CAS-Nummer: 1159976-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...