N,N-Diethyl-1,4-phenylenediamine hydrochloride

Lieferant: Apollo Scientific

OR307789-5G
APOSOR307789-5GEA 143 EUR
APOSOR307789-5G
N,N-Diethyl-1,4-phenylenediamine hydrochloride
N,N-Diethyl-1,4-phenylenediamine hydrochloride
Formel: C₁₀H₁₆N₂·ClH
Molecular Weight: 200,708 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035501
CAS-Nummer: 2198-58-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR