3-(4-Nitrobenzylamino)pyrrolidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

OR307790-1G
APOSOR307790-1GEA 285 EUR
APOSOR307790-1G
3-(4-Nitrobenzylamino)pyrrolidine-1-carboxylic acid tert-butyl ester
3-(4-Nitrobenzylamino)pyrrolidine-1-carboxylic acid tert-butyl ester
Formel: C₁₆H₂₃N₃O₄
Molecular Weight: 321,372 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12963738
CAS-Nummer: 1204811-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR