tert-Butyl 4-(4-amino-2-chlorophenyl)piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR307791-1G
APOSOR307791-1GEA 325 EUR
APOSOR307791-1G
tert-Butyl 4-(4-amino-2-chlorophenyl)piperazine-1-carboxylate
tert-Butyl 4-(4-amino-2-chlorophenyl)piperazine-1-carboxylate
Formel: C₁₅H₂₂ClN₃O₂
Molecular Weight: 311,807 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519255
CAS-Nummer: 193902-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR