tert-Butyl 4-(4-amino-2-methylphenyl)piperazine-1-carboxylate

Lieferant: Apollo Scientific

OR307792-1G
APOSOR307792-1GEA 317 EUR
APOSOR307792-1G
tert-Butyl 4-(4-amino-2-methylphenyl)piperazine-1-carboxylate
tert-Butyl 4-(4-amino-2-methylphenyl)piperazine-1-carboxylate
Formel: C₁₆H₂₅N₃O₂
Molecular Weight: 291,389 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11519254
CAS-Nummer: 361345-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR