3-Morpholin-4-ylmethyl-phenylamine dihydrochloride

Lieferant: Apollo Scientific

OR307794-1G
APOSOR307794-1GEA 119 EUR
APOSOR307794-1G
3-Morpholin-4-ylmethyl-phenylamine dihydrochloride
3-Morpholin-4-ylmethyl-phenylamine dihydrochloride
Formel: C₁₁H₁₆N₂O·2ClH
Molecular Weight: 265,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08456821
CAS-Nummer: 1431364-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR