1-(3-Aminophenyl)piperidine dihydrochloride

Lieferant: Apollo Scientific

OR307795-1G
APOSOR307795-1GEA 185 EUR
APOSOR307795-1G
1-(3-Aminophenyl)piperidine dihydrochloride
1-(3-Aminophenyl)piperidine dihydrochloride
Formel: C₁₁H₁₆N₂·2ClH
Molecular Weight: 249,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24368290
CAS-Nummer: 138227-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR