3-Pyrrolidin-1-ylmethylphenylamine

Lieferant: Apollo Scientific

Synonyme: 3-(Pyrrolidin-1-ylmethyl)aniline

OR307796-1G
APOSOR307796-1GEA 127 EUR
APOSOR307796-1G
3-Pyrrolidin-1-ylmethylphenylamine
3-Pyrrolidin-1-ylmethylphenylamine
Formel: C₁₁H₁₆N₂
Molecular Weight: 176,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114504
CAS-Nummer: 183365-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR