5,7-Dihydroxy-2,2-dimethylchroman-4-one 95%

Lieferant: Apollo Scientific
Achtung

OR307797-1G
APOSOR307797-1GEA 231 EUR
APOSOR307797-1G
5,7-Dihydroxy-2,2-dimethylchroman-4-one 95%
5,7-Dihydroxy-2,2-dimethylchroman-4-one
Formel: C₁₁H₁₂O₄
Molecular Weight: 208,211 g/mol
Schmelzpunkt: 187…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02066217
CAS-Nummer: 883-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR