10-Bromo-2,2,6,6-tetramethyl-2H-1,5,7-trioxa-phenanthren-8-one

Lieferant: Apollo Scientific

OR307799-1G
APOSOR307799-1GEA 665 EUR
APOSOR307799-1G
10-Bromo-2,2,6,6-tetramethyl-2H-1,5,7-trioxa-phenanthren-8-one
10-Bromo-2,2,6,6-tetramethyl-2H-1,5,7-trioxa-phenanthren-8-one
Formel: C₁₅H₁₅BrO₄
Molecular Weight: 339,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06409234
CAS-Nummer: 531501-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR