(1R,2R)-4-Oxo-cyclopentane-1,2-dicarboxylic acid dimethyl ester

Lieferant: Apollo Scientific
Achtung

OR307802-250MG
APOSOR307802-250MGEA 509 EUR
APOSOR307802-250MG
(1R,2R)-4-Oxo-cyclopentane-1,2-dicarboxylic acid dimethyl ester
(1R,2R)-4-Oxo-cyclopentane-1,2-dicarboxylic acid dimethyl ester
Formel: C₉H₁₂O₅
Molecular Weight: 200,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10001388
CAS-Nummer: 35079-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR