[2-[2-(2-Hydroxy-ethoxy)-ethoxy]-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester

Lieferant: Apollo Scientific
Achtung

OR307804-1G
APOSOR307804-1GEA 451 EUR
APOSOR307804-1G
[2-[2-(2-Hydroxy-ethoxy)-ethoxy]-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester
[2-[2-(2-Hydroxy-ethoxy)-ethoxy]-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester
Formel: C₂₁H₂₅NO₅
Molecular Weight: 371,427 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669697
CAS-Nummer: 560088-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR