[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: Fmoc-N-amido-PEG2-alcohol, (9H-Fluoren-9-yl)methyl (2-(2-hydroxyethoxy)ethyl)carbamate, Fmoc-5-amino-3-oxa-1-pentanol

OR307805-1G OR307805-5G
APOSOR307805-1GEA 50.9 EUR
APOSOR307805-1G APOSOR307805-5G
[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester 99%
[2-(2-Hydroxy-ethoxy)-ethyl]-carbamic acid 9H-fluoren-9-ylmethyl ester
Formel: C₁₉H₂₁NO₄
Molecular Weight: 327.374 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04974286
CAS-Nummer: 299430-87-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR