4,6-Dichloro-2-trichloromethyl-quinoline
Lieferant: Apollo Scientific
OR307815-5G
APOSOR307815-5GEA
243
EUR
APOSOR307815-5G
4,6-Dichloro-2-trichloromethyl-quinoline
4,6-Dichloro-2-trichloromethyl-quinoline
Formel:
C₁₀H₄Cl₅N Molecular Weight: 315,41 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272424 CAS-Nummer: 93600-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...