7-Bromo-2-chloroquinoline-3-carbonitrile

Lieferant: Apollo Scientific

OR307817-1G
APOSOR307817-1GEA 375 EUR
APOSOR307817-1G
7-Bromo-2-chloroquinoline-3-carbonitrile
7-Bromo-2-chloroquinoline-3-carbonitrile
Formel: C₁₀H₄BrClN₂
Molecular Weight: 267,509 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437552
CAS-Nummer: 892874-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR