7-Bromo-4-hydrazinoquinoline hydrochloride

Lieferant: Apollo Scientific

OR307819-1G OR307819-5G
APOSOR307819-1GEA 301 EUR
APOSOR307819-1G APOSOR307819-5G
7-Bromo-4-hydrazinoquinoline hydrochloride
7-Bromo-4-hydrazinoquinoline hydrochloride
Formel: C₉H₈BrN₃·ClH
Molecular Weight: 274.545 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07700271
CAS-Nummer: 1172413-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR