6-Chloro-4-hydroxy-8-methylquinoline

Lieferant: Apollo Scientific

OR307829-1G OR307829-250MG
APOSOR307829-1GEA 261 EUR
APOSOR307829-1G APOSOR307829-250MG
6-Chloro-4-hydroxy-8-methylquinoline
6-Chloro-4-hydroxy-8-methylquinoline
Formel: C₁₀H₈ClNO
Molecular Weight: 193.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272439
CAS-Nummer: 203626-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR