5-(4-Morpholinyl)-2-nitroanilin
Lieferant: Apollo Scientific
OR30783-1G
OR30783-5G
APOSOR30783-1GEA
94.4
EUR
APOSOR30783-1G
APOSOR30783-5G
5-(4-Morpholinyl)-2-nitroanilin
5-(4-Morpholinyl)-2-nitroanilin
Formel:
C₁₀H₁₃N₃O₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD00511844 CAS-Nummer: 54998-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...