7-Chlor-8-methylchinolin-4-ol

Lieferant: Apollo Scientific

Synonyme: 7-Chloro-4-hydroxy-8-methylquinoline

OR307830-5G
APOSOR307830-5GEA 134 EUR
APOSOR307830-5G
7-Chlor-8-methylchinolin-4-ol
7-Chlor-8-methylchinolin-4-ol
Formel: C₁₀H₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00272440
CAS-Nummer: 203626-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR