8-Chloroquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR307837-1G
APOSOR307837-1GEA 306 EUR
APOSOR307837-1G
8-Chloroquinoline-3-carboxylic acid
8-Chloroquinoline-3-carboxylic acid
Formel: C₁₀H₆ClNO₂
Molecular Weight: 207,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437568
CAS-Nummer: 71082-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR