8-Chloro-4-hydroxyquinoline

Lieferant: Apollo Scientific

OR307840-1G
APOSOR307840-1GEA 84.1 EUR
APOSOR307840-1G
8-Chloro-4-hydroxyquinoline
8-Chloro-4-hydroxyquinoline
Formel: C₉H₆ClNO
Molecular Weight: 179,603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272388
CAS-Nummer: 57797-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR