8-Methyl-quinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR307848-1G
APOSOR307848-1GEA 0 EUR
APOSOR307848-1G
8-Methyl-quinoline-3-carboxylic acid
8-Methyl-quinoline-3-carboxylic acid
Formel: C₁₁H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08437582
CAS-Nummer: 71082-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR