2-Methyl-4,7,8-trichlorchinolin

Lieferant: Apollo Scientific

Synonyme: 4,7,8-Trichloro-2-methylquinoline

OR307850-1G
APOSOR307850-1GEA 270 EUR
APOSOR307850-1G
2-Methyl-4,7,8-trichlorchinolin
2-Methyl-4,7,8-trichloroquinoline
Formel: C₁₀H₆Cl₃N
Molecular Weight: 246.520 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272309
CAS-Nummer: 108097-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR