2,3-Quinoline dicarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR307851-1G
APOSOR307851-1GEA 0 EUR
APOSOR307851-1G
2,3-Quinoline dicarboxylic acid
2,3-Quinoline dicarboxylic acid
Formel: C₁₁H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00071756
CAS-Nummer: 643-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR