2,6,7-Trimethylchinolin
Lieferant: Apollo Scientific
OR307852-1G
APOSOR307852-1GEA
359
EUR
APOSOR307852-1G
2,6,7-Trimethylchinolin
2,6,7-Trimethylquinoline
Formel:
C₁₂H₁₃N Lagertemperatur: Raumtemperatur |
CAS-Nummer:
72681-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...