7-Chloro-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

OR307855-1G
APOSOR307855-1GEA 293 EUR
APOSOR307855-1G
7-Chloro-4-hydroxy-2-methylquinoline
7-Chloro-4-hydroxy-2-methylquinoline
Formel: C₁₀H₈ClNO
Molecular Weight: 193.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00272320
CAS-Nummer: 15644-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR