2-Amino-6-bromoquinoline hydrochloride

Lieferant: Apollo Scientific

OR307864-1G
APOSOR307864-1GEA 301 EUR
APOSOR307864-1G
2-Amino-6-bromoquinoline hydrochloride
2-Amino-6-bromoquinoline hydrochloride
Formel: C₉H₇BrN₂·ClH
Molecular Weight: 259,53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703346
CAS-Nummer: 1170935-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR