6-Methoxyquinoline-3-carboxaldehyde

Lieferant: Apollo Scientific

OR307866-1G
APOSOR307866-1GEA 565 EUR
APOSOR307866-1G
6-Methoxyquinoline-3-carboxaldehyde
6-Methoxyquinoline-3-carboxaldehyde
Formel: C₁₁H₉NO₂
Molecular Weight: 187,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09260427
CAS-Nummer: 13669-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR