6-Bromo-4-chloro-2-isopropylquinoline hydrochloride

Lieferant: Apollo Scientific

OR307873-1G
APOSOR307873-1GEA 517 EUR
APOSOR307873-1G
6-Bromo-4-chloro-2-isopropylquinoline hydrochloride
6-Bromo-4-chloro-2-isopropylquinoline hydrochloride
Formel: C₁₂H₁₁BrClN·ClH
Molecular Weight: 321,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09475440
CAS-Nummer: 1170370-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR