6-Brom-4-chlor-2-phenylchinolin

Lieferant: Apollo Scientific

OR307875-1G
APOSOR307875-1GEA 383 EUR
APOSOR307875-1G
6-Brom-4-chlor-2-phenylchinolin
6-Bromo-4-chloro-2-phenylquinoline
Formel: C₁₅H₉BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD02684146
CAS-Nummer: 860195-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR