2,5,7-Trimethyl-3-chinolincarbonsäureethylester

Lieferant: Apollo Scientific

OR307878-1G OR307878-5G
APOSOR307878-1GEA 293 EUR
APOSOR307878-1G APOSOR307878-5G
2,5,7-Trimethyl-3-chinolincarbonsäureethylester
2,5,7-Trimethylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₅H₁₇NO₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 948291-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR