2,5-Dichloro-8-methylquinoline-3-carbonitrile

Lieferant: Apollo Scientific

OR307880-1G
APOSOR307880-1GEA 375 EUR
APOSOR307880-1G
2,5-Dichloro-8-methylquinoline-3-carbonitrile
2,5-Dichloro-8-methylquinoline-3-carbonitrile
Formel: C₁₁H₆Cl₂N₂
Molecular Weight: 237,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787536
CAS-Nummer: 948291-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR