2,5-Dichloro-8-methylquinoline-3-carboxaldehyde

Lieferant: Apollo Scientific

OR307881-1G
APOSOR307881-1GEA 383 EUR
APOSOR307881-1G
2,5-Dichloro-8-methylquinoline-3-carboxaldehyde
2,5-Dichloro-8-methylquinoline-3-carboxaldehyde
Formel: C₁₁H₇Cl₂NO
Molecular Weight: 240,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787537
CAS-Nummer: 948291-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR