2,6-Dichlor-3-methylchinolin

Lieferant: Apollo Scientific
Achtung

OR307884-1G
APOSOR307884-1GEA 255 EUR
APOSOR307884-1G
2,6-Dichlor-3-methylchinolin
2,6-Dichloro-3-methylquinoline
Formel: C₁₀H₇Cl₂N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 132118-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR