2,6-Dichloro-8-methylquinoline-3-carbonitrile

Lieferant: Apollo Scientific

OR307885-1G
APOSOR307885-1GEA 375 EUR
APOSOR307885-1G
2,6-Dichloro-8-methylquinoline-3-carbonitrile
2,6-Dichloro-8-methylquinoline-3-carbonitrile
Formel: C₁₁H₆Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09787542
CAS-Nummer: 948291-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR