2-Amino-6-bromo-3-phenylquinoline hydrochloride

Lieferant: Apollo Scientific

OR307892-1G
APOSOR307892-1GEA 598 EUR
APOSOR307892-1G
2-Amino-6-bromo-3-phenylquinoline hydrochloride
2-Amino-6-bromo-3-phenylquinoline hydrochloride
Formel: C₁₅H₁₁BrN₂·ClH
Molecular Weight: 335,626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703381
CAS-Nummer: 1170960-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR