2-Amino-6-chloro-8-methyl-3-phenylquinoline hydrochloride

Lieferant: Apollo Scientific

OR307893-1G
APOSOR307893-1GEA 359 EUR
APOSOR307893-1G
2-Amino-6-chloro-8-methyl-3-phenylquinoline hydrochloride
2-Amino-6-chloro-8-methyl-3-phenylquinoline hydrochloride
Formel: C₁₆H₁₃ClN₂·ClH
Molecular Weight: 305,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703388
CAS-Nummer: 1171355-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR