2-Chloro-3-(2-chloroethyl)-6-bromoquinoline

Lieferant: Apollo Scientific

OR307894-1G
APOSOR307894-1GEA 351 EUR
APOSOR307894-1G
2-Chloro-3-(2-chloroethyl)-6-bromoquinoline
2-Chloro-3-(2-chloroethyl)-6-bromoquinoline
Formel: C₁₁H₈BrCl₂N
Molecular Weight: 304.998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09787617
CAS-Nummer: 948294-46-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR